Drucken

Anti-IgG Goat Polyclonal Antibody (CF™ 750)

Lieferant: Biotium
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

20070 20070-1
20070.EA 327 EUR
20070. 20070-1
Anti-IgG Goat Polyclonal Antibody (CF™ 750)
Antikörper
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 750 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR