Drucken

Anti-IgG Goat Polyclonal Antibody (CF™ 568)

Lieferant: Biotium
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

20108 20108-1
20108.EA 300 EUR
20108. 20108-1
Anti-IgG Goat Polyclonal Antibody (CF™ 568)
Antikörper
Type: Secondary
Antigen: IgG (Guinea pig) (guinea pig IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 568 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Guinea pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR