Drucken

Anti-IgG Goat Polyclonal Antibody (CF™ 633)

Lieferant: Biotium
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

20126 20126-1
20126.EA 359 EUR
20126. 20126-1
Anti-IgG Goat Polyclonal Antibody (CF™ 633)
Antikörper
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR