Drucken
Follicle dermal papilla cell culture medium
Lieferant: PromoCell
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
C-26500EA 225 EUR
C-26500 C-26505 C-26502 C-26501
Follicle dermal papilla cell culture medium
Zellkulturmedien
PromoCell Follicle Dermal Papilla Cell Culture Medium is a low-serum medium. The combination of low serum and growth factors leads to excellent proliferation of human follicle dermal papilla cells in vitro.
Lieferumfang: The medium (ready-to-use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix. Medium Kit consists of 1× 500 ml bottle basal medium and 1× SupplementPack (individual vials with pre-measured supplements).