Drucken
Hematopoietic Progenitor Medium
Lieferant: PromoCell
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
POMOC-28020EA 85 EUR
POMOC-28020
Hematopoietic Progenitor Medium
PromoCell Hematopoietic Progenitor Medium was developed for the expansion of hematopoietic progenitors by means of differentiation into more committed lineages of blood cells. Hence, this progenitor medium is suitable for CFU-based assays.