Drucken

Anti-ANGPTL6 Mouse Monoclonal Antibody [clone: Kairos-60]

Lieferant: Cayman Chemical
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

CAYM10808-100EA 330 EUR
CAYM10808-100 CAYM10808-50
Anti-ANGPTL6 Mouse Monoclonal Antibody [clone: Kairos-60]
Type: Primary
Antigen: ANGPTL6
Clonality: Monoclonal
Clone: Kairos-60
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR