Drucken

Anti-eIF3G Rabbit Polyclonal Antibody

Lieferant: Cayman Chemical
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

CAYM13537-1EA 332 EUR
CAYM13537-1
Anti-eIF3G Rabbit Polyclonal Antibody
Type: Primary
Antigen: eIF3G
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Chimpanzee, Horse, Human, Mouse, Rat, Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR