Drucken

Anti-AcK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Cayman Chemical
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

CAYM13726-400EA 589 EUR
CAYM13726-400
Anti-AcK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Primary
Antigen: AcK
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish PE (Phycoerythrin)roxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: multi-species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR