Drucken
Metal block banks for paraffin blocks
Lieferant: Leica
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
LEIH3808550EEA 549 EUR
LEIH3808550E
Metal block banks for paraffin blocks
Möbel Lagerschränke
These block banks are ideal for the long-term storage of paraffin blocks. These rigid steel cabinets will stack together or with the slide safe.
  • Available with matching tops, black base plinth or wheeled plinth
  • Capacity up to 3000 blocks