Drucken

Anti-HA Mouse Monoclonal Antibody (RPE (R-Phycoerythrin))

Lieferant: Immunoreagents
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

IMMRIR2833EA 469 EUR
IMMRIR2833
Anti-HA Mouse Monoclonal Antibody (RPE (R-Phycoerythrin))
R-phycoerythrin is used in immunoassays such as FACS, flow cytometry, multimer/tetramer applications. With new instrumetation available, R-PE is also well suited for immunohistochemistry (IHC) and Western blot applications.

Type: Tag
Antigen: HA
Clonality: Monoclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR