Drucken

Anti-S-Tag Antibody (DyLight® 488)

Lieferant: Immunoreagents
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

IMMRIR2846EA 469 EUR
IMMRIR2846
Anti-S-Tag Antibody (DyLight® 488)
The ImmunoReagents' DyLight® 488 conjugates provide exceptional performance for a wide array of fluorescence-base applications at a lower cost than other 488-range dyes. DyLight® 488 shows greater photostability than traditional fluorescein-based dyes and has a greater range of pH insensitivity (pH 4-9). DyLight® 488 conjugates are available for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 493nm / 518nm Emission Color = Green (Similar Dyes: Alexa Fluor 488, Cy2, FITC)

Type: Tag
Antigen: S-Tag
Clonality:
Clone:
Conjugation: DyLight® 488
Epitope:
Host:
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR