Drucken

Anti-UBXD2 Rabbit Polyclonal Antibody

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

BOSSBS-11675REA 691 EUR
BOSSBS-11675R
Anti-UBXD2 Rabbit Polyclonal Antibody
Antikörper
Involved in endoplasmic reticulum-associated protein degradation (ERAD). Acts as a platform to recruit both UBQLN1 and VCP to the ER during ERAD (PubMed:19822669).

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: UBXD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR