Graphite, nickel coated (≥99,8% Ni) (Metall-Basis), Pulver -100 mesh
Lieferant: ALFA AESAR
Ratings:
(keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
|
Gefahr
|
43215.22
43215.36
43215.22EA
49.4
EUR
43215.22
43215.36
Graphite, nickel coated (≥99,8% Ni) (Metall-Basis), Pulver -100 mesh
Graphite, nickel coated
Purity: 99.8%
Physical Form: Ni:C; 60:40 wt%
Appearance: Black
Odour: Odourless
UN No.: UN3178
MDL No.: MFCD00144065
Applications: Conductive surface coatings, inks, adhesives, and sealants
Storage Temperature:
Raumtemperatur |
MDL Number:
MFCD00144065 UN: 3178 ADR: 4.1 III |