Drucken

Transformation reagent, LiSorb

Lieferant: Thermo Fisher Scientific
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

J60087.APEA 39.6 EUR
ALFAJ60087.AP ALFAJ60087.AK
Transformation reagent, LiSorb
For transformation of competent cells.

Physical Form: Liquid

Applications: For transformation of competent cells
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR