Drucken
Hybrid refractometers and polarimeters
Lieferant: Atago
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
ATAG5012EA 2410 EUR
ATAG5012 ATAG5013 ATAG5014 ATAG5010 ATAG5015
Hybrid refractometers and polarimeters
This refractometer/polarimeter is designed for use in the sugar manufacturing, refineries, fragrances and honey analysis.
  • Green and red light alarms
  • Selectable display unit of measure via display
  • Automatic power-off when the instrument is not in use
  • High accuracy (Brix and optical rotation) from one sample
  • Compact size saves bench space by combining two instruments in one
Zero the unit with distilled water in order to calibrate measurements.
Lieferumfang: Supplied with four alcaline batteries.