Drucken
Laboratory refrigerators, GPS series
Lieferant: Thermo Scientific
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
HERAR14X-GAEW-TSEA 3662 EUR
HERAR14X-GAEW-TS
Laboratory refrigerators, GPS series
Protect routine samples with lab refrigerators designed for general purpose lab storage.
  • Integrated controller
  • Digital display and with access ports
  • High and low temperature alarms
  • Low energy consumption
Zertifizierungen: CE marked