Drucken
Immersed body (Plummet)
Lieferant: Industrial Physics
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
SIMEDV5000EA 871 EUR
SIMEDV5000
Immersed body (Plummet)
The immersed body (260 mm) is designed to determine the density of paints or related products at 23 °C.
The method is based on 'Archimedes' principle.
The upward force caused by immersing the plummet into the liquid is read from a laboratory balance and expressed in grams per milliliter.
Zertifizierungen: DIN EN ISO 2811-2
Lieferumfang: Supplied in plastic case - A stand needs to be ordered separately.