Drucken
Anti-PAX7 Mouse Monoclonal Antibody (CF543) [clone: PAX7/497]
Lieferant: Biotium
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
BNC430497-500 BNC430497-100
BNC430497-500EA 560 EUR
BNC430497-500 BNC430497-100
Anti-PAX7 Mouse Monoclonal Antibody (CF543) [clone: PAX7/497]
Antikörper
The Pax gene family of nuclear transcription factors is comprised of nine members that function during embryogenesis to regulate the temporal and position-dependent differentiation of cells. In addition, the family is involved in a variety of signal transduction pathways in the adult organism. Mutations in the Pax family of proteins have been linked to disease and cancer in humans. Pax-7 is a protein specifically expressed in cultured satellite cell-derived myoblasts. In situ hybridization reveals that Pax-7 is also expressed in satellite cells residing in adult muscle. A chromosomal aberration in the gene encoding Pax-7 causes rhabdomyosarcoma 2 (RMS2) (also called alveolar rhabdomyosarcoma).
Type: Primary
Antigen: PAX7
Clonality: Monoclonal
Clone: PAX7/497
Conjugation: CF543
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chicken, Human, Mouse, Rat, Zebrafish