Drucken
Anti-Ep-CAM Mouse Monoclonal Antibody (CF647) [clone: EGP40/837]
Lieferant: Biotium
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
BNC470837-500 BNC470837-100
BNC470837-500EA 560 EUR
BNC470837-500 BNC470837-100
Anti-Ep-CAM Mouse Monoclonal Antibody (CF647) [clone: EGP40/837]
Antikörper
EGP40 is a 40-43 kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas with the exception of adult squamous epithelium, hepatocytes and gastric epithelial cells. This antibody has been used to distinguish adenocarcinoma from pleural mesothelioma and hepatocellular carcinoma. This antibody is also useful in distinguishing serous carcinomas of the ovary from mesothelioma.
Type: Primary
Antigen: Ep-CAM
Clonality: Monoclonal
Clone: EGP40/837
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human