Drucken
Anti-CD8A Mouse Monoclonal Antibody (CF568) [clone: C8/1035]
Lieferant: Biotium
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
BNC681035-500 BNC681035-100
BNC681035-500EA 560 EUR
BNC681035-500 BNC681035-100
Anti-CD8A Mouse Monoclonal Antibody (CF568) [clone: C8/1035]
Antikörper
CD8 is a cell surface receptor expressed either as a heterodimer with the CD8 β chain (CD8 α/β) or as a homodimer (CD8 α/α). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class 1 and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells. For mature T-cells, CD4 and CD8 are mutually exclusive, so anti-CD8, generally used in conjunction with anti-CD4. It is a useful marker for distinguishing helper/inducer T-lymphocytes, and most peripheral T-cell lymphomas are CD4 /CD8-. Anaplastic large cell lymphoma is usually CD4 and CD8-, and in T-lymphoblastic lymphoma/leukemia, CD4 and CD8 are often co-expressed. CD8 is also found in littoral cell angioma of the spleen.
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: C8/1035
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human