Drucken
Anti-BrdU Mouse Monoclonal Antibody (Purified) [clone: BRD469]
Lieferant: Biotium
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
BNUB0469-100 BNUB0469-500
BNUB0469-100EA 237 EUR
BNUB0469-100 BNUB0469-500
Anti-BrdU Mouse Monoclonal Antibody (Purified) [clone: BRD469]
Antikörper
This antibody reacts with Bromodeoxyuridine (BrdU) in single stranded DNA (produced by partial denaturation of double stranded DNA), BrdU coupled to a protein carrier, as well as free BrdU. BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.
Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: BRD469
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All species