Drucken
Membrane filters, type 131
Lieferant: Sartorius
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
SART13101-50-AHNEA 218 EUR
SART13101-50-AHN
Membrane filters, type 131
Filter Membranfilter
Non sterile membrane filters for colony counting, particle testing, microscopy and sterility testing. The membrane filters are made of cellulose nitrate (cellulose ester).
  • High flow membranes