Drucken
Labels for TLS 2200® and TLS PC Link™ printers, type B-461
Lieferant: Brady
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
062255 062258
BRDY062255EA 126.86 EUR
BRDY062255 BRDY062258
Labels for TLS 2200® and TLS PC Link™ printers, type B-461
Etiketten
These TLS2200® & TLS PC Link™ labels are ideal for use with thermal transfer portable printers.
  • Self-laminating
  • Excellent abrasion and smudge resistance
TLS2200® parts can also be printed on the BMP™71 label printer provided the roll adapter (M71-ADAPT) is used.