Drucken
Autosampler moulding kit
Lieferant: Perkin Elmer
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
PERCL1245025EA 0 EUR
PERCL1245025
Autosampler moulding kit
Formen Formen für Autosampler
The autosampler moulding kit consist of all parts required for the creation of more than hundred custom moulds.
  • Reusable for measurements up to 100 times
  • Resin has a shelf life of six months
  • With tablet setting pins 10 moulds can be routinely produced
Bestellinformation: Pins can be ordered separately to increase mould formation throughput.