Drucken

Anti-SDK2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

BOSSBS-11165R-CY5EA 425 EUR
BOSSBS-11165R-CY5
Anti-SDK2 Rabbit Polyclonal Antibody (Cy5®)
Sidekick-2 (SDK2), also known as KIAA1514, is a 2,172 amino acid single-pass type I membrane protein containing thirteen fibronectin type-III domains and six Ig-like C2-type (immunoglobulin-like) domains. Acting as a cell adhesion protein, Sidekick-2 guides axonal terminals to synapses in developing neurons. Sidekick-2 may require the interaction with MAGI scaffolding proteins for proper localization and function. Existing as four alternatively spliced isoforms, the gene encoding Sidekick-2 maps to human chromosome 17q25.1. Chromosome 17 makes up over 2.5% of the human genome with about 81 million bases encoding over 1,200 genes. Neurofibromatosis, a condition characterized by neural and epidermal lesions, and dysregulated Schwann cell growth have been linked to chromosome 17. Alexander disease, Birt-Hogg-Dube syndrome and Canavan disease are also associated with chromosome 17.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SDK2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR