Drucken

Anti-PRKDC Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

BOSSBS-2092R-CY5.5EA 437 EUR
BOSSBS-2092R-CY5.5
Anti-PRKDC Rabbit Polyclonal Antibody (Cy5.5®)
DNA PK is the key component of the non-homologous end-joining (NHEJ) pathway of DSB repair in mammalian cells. DNA PK consists of a heterodimeric DNA-binding subunit (Ku70/80) and an approximately 465 kDa catalytic subunit (DNA PKcs). DNA PKcs is a serine/threonine protein kinase whose activity is greatly stimulated by its recruitment to DNA breaks by the Ku heterodimer.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PRKDC
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR