Drucken

Anti-ABP1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

BOSSBS-4733R-A350EA 437 EUR
BOSSBS-4733R-A350
Anti-ABP1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
ABP1

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR