Drucken
PLOT columns, BP BOND Q
Lieferant: SGE (Trajan)
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
SGEA0570535EA 762 EUR
SGEA0570535 SGEA0570123 SGEA0570223 SGEA0570135 SGEA0570235
PLOT columns, BP BOND Q
Highly stable columns for non-polar and semi-volatile compounds.
  • Withstand repeated water injections
  • Reduced need for particle trap due to minimal particle shredding
  • Broad application range - ideal for volatile solvents and hydrocarbon analysis
PLOT columns are ideal for separating compounds that are gases at room temperature. Trajan Scientific and Medical offers the BP BOND range of PLOT columns for analysis of fixed gases, low molecular weight hydrocarbons, volatile polar compounds and reactive analytes such as sulphur gases, amines and hydrides. BP BOND PLOT columns are engineered to prevent the stationary phase particles from shredding downstream. Columns have been manufactured for stability at high temperatures, low bleed and high inertness levels.