Drucken
General purpose gloves, Abratex Light
Lieferant: Honeywell Safety Products
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
DALLRGT088/6EA 27.1 EUR
DALLRGT088/6 DALLRGT088/9 DALLRGT088/10
General purpose gloves, Abratex Light
These knitted gloves are made from 67% grey polyamid and 33% cotton.
  • Knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
  • Cotton interior slows down perspiration and increases comfort
  • Blend of two fibres (polyamide and cotton) for better abrasion and cutting resistance
  • White colour enhances the glove's hygiene and cleanliness properties
  • Knitted wrist (5 cm) with size related coloured edge
Ideal for light handlings of fine parts and assembling.
Zertifizierungen: EN 388:2016 (224XX); Cat. II