Drucken
T4 DNA Ligase
Lieferant: Novagene
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
69839-3EA 72.3 EUR
NOVG69839-3
T4 DNA Ligase
T4 DNA ligase catalyses the formation of phosphodiester bonds between 3'-hydroxyl and 5'-phosphate groups of adjacent DNA and RNA nucleotides in blunt-end or cohesive-end configurations. The enzyme joins RNA and DNA strands only in duplex molecules and does not join single-stranded nucleic acids.
These are rigorously tested in a blue-white cloning assay under conditions that maximise exposure of DNA ends to any contaminating nucleases. The enzyme is qualified for any ligation or cloning application.