Drucken

Anti-Annexin V Rabbit Polyclonal Antibody

Lieferant: PromoCell
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

PK-AB577-3357EA 369 EUR
PK-AB577-3357
Anti-Annexin V Rabbit Polyclonal Antibody
Rabbit Anti-Human/Mouse/Rat Annexin V

Type: Primary
Antigen: Annexin V
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: The antibody detects Annexin V from samples of human, mouse, and rat origin.
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR