Drucken

Anti-MICA Rabbit Polyclonal Antibody

Lieferant: PromoCell
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

PK-AB718-4261EA 359 EUR
PK-AB718-4261
Anti-MICA Rabbit Polyclonal Antibody
MICA antibody can be used for detection of MICA by Western blot at 0.5 - 2 µg/mL. Antibody can also be used for immunohistochemistry starting at 10 µg/mL. For immunofluorescence start at 20 µg/mL.

Type: Primary
Antigen: MICA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR