Drucken

Anti-Unc93a Rabbit Polyclonal Antibody

Lieferant: PromoCell
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

PK-AB718-4551EA 359 EUR
PK-AB718-4551
Anti-Unc93a Rabbit Polyclonal Antibody
Unc93a antibody can be used for the detection of Unc93a by Western blot at 0.5 - 1 µg/mL.

Type: Primary
Antigen: Unc93a
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR