Drucken
L-Lactate Assay Kit II
Lieferant: PromoKine
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
POMOPK-CA577-K607EA 489 EUR
POMOPK-CA577-K607
L-Lactate Assay Kit II
The L-Lactate Assay Kit II provides a convenient means for detecting L(+)-Lactate in various biological samples.
  • No need of pre-treatment or purification of samples
  • Lactate specifically reacts with an enzyme mix
In the fluorometric/colorimetric L-Lactate Assay Kit II, lactate specifically reacts with an enzyme mix to generate a product, which interacts with a lactate probe to produce a signal detectable as a colour (570 nm) or as fluorescence (Ex/Em = 535/587 nm). The kit provides a convenient means for detecting L(+)-Lactate in various biological samples, e.g. in circulating blood, cells, culture media, fermentation media, etc. at concentrations between 0,001 to 10 mM. There is no need of pre-treatment or purification of samples.