Drucken
HPLC columns, InfinityLab Poroshell 120 HILIC
Lieferant: Agilent
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
683775-601 685775-601 683975-601 685975-601 683775-924 683975-324 683975-924 685775-924 685975-324 685975-924 820750-933 821725-947 823750-948
HEWL683775-601EA 1276 EUR
HEWL683775-601 HEWL685775-601 HEWL683975-601 HEWL685975-601 HEWL683775-924 HEWL683975-324 HEWL683975-924 HEWL685775-924 HEWL685975-324 HEWL685975-924 HEWL820750-933 HEWL821725-947 HEWL823750-948
HPLC columns, InfinityLab Poroshell 120 HILIC
Chromatographiesäulen
InfinityLab Poroshell 120 HILIC chemistries combine innovative particle technology and bonded phases to improve retention and peak shape for polar analytes. This enables quick and efficient separations of challenging, highly charged polar compounds, and achieves superior analytical sensitivity in MS mode using MS-friendly eluents.
InfinityLab Poroshell 120 HILIC-Z columns:
• Novel zwitterionic stationary phase bonded to 2,7 µm Poroshell 120 particles
• PEEK-lined column option for excellent peak shape and recovery of particularly challenging charged compounds
• High pH and temperature stability: Up to pH 12 and 80 ºC
InfinityLab Poroshell 120 HILIC-OH5 columns:
• Novel poly-hydroxy fructan phase bonded to 2,7 µm Poroshell 120 particles
• Alternate selectivity to HILIC and HILIC-Z phases