Drucken
Gardner colour reference standards, ASTM D 1544, D 6166, Supelco®
Lieferant: Merck
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
SIAL134200-500MLEA 175 EUR
SIAL134200-500ML SIAL134220-500ML SIAL134210-500ML
Gardner colour reference standards, ASTM D 1544, D 6166, Supelco®
Gardner Colour Standards are for routine calibration of color measuring instruments and verification of test data. Process industries throughout the world use colorimeters for color control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints, and coatings.
Zertifizierungen: This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 (ISO Guide 34) under UKAS accreditation.