Drucken
HyClone™ Fetal Bovine Serum, Canadian Origin, HyClone Products (Cytiva)
Lieferant: HyClone products (Cytiva)
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
HyClone™
HYCLSH30396.02EA 175 EUR
HYCLSH30396.02 HYCLSH30396.03
HyClone™ Fetal Bovine Serum, Canadian Origin, HyClone Products (Cytiva)
Seren
HyClone Characterized Fetal Bovine Serum, Canadian origin, uses "True Pool" processing technology to ensure consistency in the quality of serum.
  • Assayed for a variety of critical components and biochemical characteristics
  • Endotoxin-tested: certificate of analysis confirms ≤25 EU/ml endotoxin
  • Post-filtration treatments
The final serum is assayed for provides important proteins, hormones, growth factors, metabolites, and nutrients for that support your healthy cell culture process. Assay results are included in the certificate of analysis and biochemical assay list.
Additional testing, including EMEA conformance testing, is also available.
Gamma-irradiated and heat-inactivated FBS options are available to further reduce the risk of adventitious agents, particularly when manufacturing products for use in humans or animals. The irradiation process is validated to deliver a minimum dose of 25 kGy.