Drucken
Mammalian transfection kit
Lieferant: Agilent Technologies
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
AGLS200385EA 217 EUR
AGLS200385
Mammalian transfection kit
The mammalian transfection kits provide all the necessary reagents in a simplified protocol to efficiently and rapidly conduct DNA transfection in mammalian cells.
  • Up to 100-fold higher yield of stably infected colonies than standard CaPO₄ based techniques
  • Achieve high efficiency transfections in a variety of cell lines
For high-efficiency transfection of exogenous genes into mammalian cells in tissue culture.
Lieferumfang: Contains: Transfection buffers, Control plasmid vector: pWLneo: G418 (neomycin) selection.
Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.