Drucken
Masterflex® PVC Pressure Relief Valves, Avantor®
Lieferant: Avantor Fluid Handling
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Masterflex®
MFLX00918-50EA 0 EUR
MFLX00918-50
Masterflex® PVC Pressure Relief Valves, Avantor®
Ventile Überdruckventile
Easily meter fluids with pressure relief valves.
  • Choose from different max pressures based on your application
These PVC valves have O-rings of Viton®, making them Ideal for liquid applications.