Drucken
Hayward® Vacuum Breaker Vent Valves, PVC
Lieferant: Hayward
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
MFLX06225-95EA 241.88 EUR
MFLX06225-95
Hayward® Vacuum Breaker Vent Valves, PVC
Ventile Luft-Regelventile
Eliminate problems when evacuating closed tanks or piping systems.
  • Constructed of durable PVC with a Viton® seal
Eliminates problems when evacuating closed tanks or piping systems. The one-way valve that is normally closed at positive pressure will allow drainage at a rate of 300 GPM. For higher drainage rates use additional vacuum breakers. The 3⁄4" NPT(F)socket adapts easily onto a 11⁄2" bulkhead fitting. The breaker is made of PVC with a Viton® seal.