Drucken
Rechargeable Electric Backpack Sprayer
Lieferant: Avantor Fluid Handling
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
MFLX70605-05EA 554.19 EUR
MFLX70605-05
Rechargeable Electric Backpack Sprayer
Sprühgeräte
Microprocessor pump control with adjustable spray tip – delicate mist, high volume, or long-range stream.
  • Tank capacity of 4 gallons
  • Comfortable ergonomic design with a full back pad and thickly padded shoulder harness.
  • Battery pack charges while in place or can be removed for remote charging.
Lightweight polyethylene tank is engineered to match the body's contour. Hand lever provides on-demand operation — depress to begin spraying and release to stop. Hand lever may also be locked in the open position to reduce operator fatigue during longer operation. Microprocessor regulates pump speed for consistent spraying at all four pump settings. High settings are great for a dense spray to cover large areas while low speeds are ideal to obtain a light mist for delicate work. A status lamp indicates when the power is on and blinks when the battery needs recharging. Battery life is 1000 cycles.
What's included: pump, tank with lid, 12 VDC battery, battery charger, 24'L spray wand, spray hose, inlet strainer, harness, and waist belt.