Drucken
Experimentation kits, sequencing the human genome
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
EDVO339EA 95.6 EUR
EDVO339
Experimentation kits, sequencing the human genome
Actual data representing important genes from automated DNA Sequencers are provided. Students determine the DNA sequence, compare and extrapolate database information and identify the gene product and other closely related proteins. Data is discussed with in the framework of the Human Genome Project.
  • For 10 lab groups
  • Complete in 45 minutes
Also required: Access to the internet.
Lieferumfang: Kit includes instructions and automated sequencing printouts.