Drucken
Anti-PD-L1 Mouse Monoclonal Antibody [clone: PDL1/2741] (CF®488A)
Lieferant: Biotium
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
BNC882741-100 BNC882741-500
BNC882741-100EA 177 EUR
BNC882741-100 BNC882741-500
Anti-PD-L1 Mouse Monoclonal Antibody [clone: PDL1/2741] (CF®488A)
Antikörper
PD-L1 is a checkpoint regulator in immune cells, it is expressed on immune or non-hematopoietic cells.
Expression of the protein is seen during pregnancy where it has a role in suppressing the immune system. PD-L1 induces an inhibitory signal in activated T-cells and promotes T-cell apoptosis. It is overexpressed in a number of different cancers where it is believed to play a significant role in the cancer s ability to evade the immune system.
Antibody/conjugate formulation: Conjugates: 0,1 mg/ml in PBS/0,1% BSA/0,05% azide, HRP conjugates: 0,1 mg/ml in PBS/0,05% BSA, Purified: 0,2 mg/ml in PBS/0,05% BSA/0,05% azide, Purified, BSA-free: 1 mg/ml in PBS without azide.
Achtung! Bitte beachten: For research use only (RUO).