Drucken
Syringe dispenser
Lieferant: Greiner bio-one
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
392-0091EA 875 EUR
392-0091
Syringe dispenser
This micro-dispenser reduces the work required to dispense eosin and formalin solutions on 60-well micro-plates. It enables reagents to be added in six wells at the same time.
  • 1 ml micro syringe used as an aspirator
  • With each press of the dosing button, 20 µl are dispensed on the six output channels, as a deposit of 3.3 µl in each well