Drucken
620-b-0024
Lieferant: Amarell
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
620-2121EA 50.57 EUR
AMARE910560
620-b-0024
These accurate thermometers are suitable for use in industry, labs, research and education.
  • Very short response time
Stainless steel probe (L×Ø: 30×2,5 mm)
Lieferumfang: Supplied with battery.