The CD 100 has a simple colour change indication as to water purity, showing when the cartridge needs changing. The unit is perfect for laboratories requiring a low cost supply of small volumes of deionised water.
- Assured water quality: a distinctive change in resin colour indicates water quality and when the cartridge needs changing
- Instant pure water anywhere, simply connect to any tap wherever pure water is needed
- Easy to use, changing cartridge takes seconds
Lieferumfang: Supplied as a housing with inlet hose and outlet spout, and with a bracket to enable wall mounting. An adapter on the inlet hose allows the unit to be connected to most taps. Supplied with first cartridge (Cat. No. 171-0423).