Drucken
Allzweckhandschuhe, Tricoton Ultralight
Lieferant: HONEYWELL SAFETY
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
RGT350/8 RGT350/9 RGT350/6 RGT350/7
DALLRGT350/8EA 18.8 EUR
DALLRGT350/8 DALLRGT350/9 DALLRGT350/6 DALLRGT350/7
Allzweckhandschuhe, Tricoton Ultralight
Handschuhe
These lightweight cotton knitted gloves limits the penetration of dirt while offering protection against small cuts.
  • Knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
  • Maximum comfort and added sturdiness
  • Pure cotton has a neutral pH level: skin-friendly, limits the absorption of perspiration
  • Light colour enhances the glove's hygiene and cleanliness properties
Ideal for fine handlings in clean environments. Can also be used as undergloves.
Zertifizierungen: Kat. I.