cAMP and cGMP conjugates
Lieferant: BIOTIUM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
00020-1 00020 00021-1 00021 00022-1 00022
00020-1EA 619 EUR InStock
00020-1 00020. 00021-1 00021. 00022-1 00022.
cAMP and cGMP conjugates
Tests Immunologische Tests
Conjugates for probing cAMP receptors or measuring cGMP in immunoassays.
  • Biotin-cAMP can be used to probe cAMP receptors
  • Biotin-cGMP has been used as a tracer in immunoassays for cGMP measurement in plasma or urine samples
  • Cyanine 644-cAMP can be used to probe cAMP receptors
Biotin-cAMP, diisopropylethylammonium salt is a biotin-conjugated cAMP analogue that can be used to probe cAMP receptors.
Biotin-cGMP, diisopropylethylammonium salt has been used as a tracer in immunoassays for the cGMP measurement in plasma or urine samples. Cyanine 644-cAMP is a fluorescently labeled cAMP analogue and can be used to probe cAMP receptors.
Cyanine 644-cAMP λExEm (MeOH) = 644/670 nm. Cyanine 644-cAMP is a fluorescently labeled cAMP analog and can be used to probe cAMP receptors. Single molecules of fluorescently labeled cAMP have been imaged as the probe binds to the receptor on the surface of Dictyostelium cells.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c71b89f5bb9fc271307593b4b8ef5e991a2f04b3&vid=c71b89f5bb9fc271307593b4b8ef5e991a2f04b3&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ea0d4df8385fb9502b1d494a8939666151f73dc&vid=3ea0d4df8385fb9502b1d494a8939666151f73dc&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82c2abf8119929b0d22bc20795e78d2ec8e3bbf8&vid=82c2abf8119929b0d22bc20795e78d2ec8e3bbf8&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge