Nitric oxide reagents and kits
Lieferant: BIOTIUM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
00242 00243 00241 00239 00249 00221-1 00221 00222 00223 00224 30100
00242.EA 212 EUR InStock
00242. 00243. 00241. 00239. 00249. 00221-1 00221. 00222. 00223. 00224. 30100.
Nitric oxide reagents and kits
Tests Zelltests
Nitric oxide donors, nitric oxide synthase inhibitors and nitric oxide detection reagents and kits for nitric oxide (NO) studies.
  • Nitric oxide donors are available as stand-alone reagents and combination kit
  • Nitric oxide synthase inhibitors are available as stand-alone reagents and combination kit
  • Nitric oxide detection kit is based on Griess reagent
Nitric Oxide Donors: SIN-1 releases nitric oxide and forms superoxide spontaneously under physiological conditions and has been shown to be a potent vasodilator. SNAP releases nitric oxide under physiological conditions and also been shown to be a potent vasodilator. Similar to SNAP, S-nitrosogluthathione releases nitric oxide spontaneously under physiological conditions. Spermine NONOate releases nitric oxide spontaneously under physiological conditions, and is useful for reliable NO generation in solution. DEA-NONOate releases nitric oxide spontaneously under physiological conditions. Useful for reliable NO generation in solution. The Nitric Oxide Generation Kit contains10 mg each of the following nitric oxide donors: SIN-1, SNAP, S-Nitrosoglutathione, spermine NONOate, and DEA-NONOate.
Nitric Oxide Synthase Inhibitors: L-NMMA competitively inhibits nitric oxide synthase in a wide range of cells. L-NIL is a potent and selective inhibitor of nitric oxide synthase that exhibits about 28-fold greater selectivity for inducible nitric oxide synthase (IC50 = 3,3 µM) than for the rat brain constitutive enzyme (IC50 = 92 µM). L-NIO is a potent, irreversible inhibitor of neutrophil nitric oxide synthase. The NOS Inhibitor kit contains 7-NI, L-NMMA, L-NIL and L-NIO.
Nitric Oxide Detection Kit: Griess reagent is used to detect nitrite photometrically. The reagent contains two chemicals, sulfanilic acid andN-(1-naphthalenediamine). Under acidic conditions sulfanilic acid is converted by nitrite to a diazonium salt, which readily couples with N-(1-naphthalenediamine) to form a highly colored azo dye that can be detected at 548 nm. Under physiological conditionss, NO is unstable and is rapidly oxidized to a mixture of nitrite and nitrate. In order to measure NO levels indirectly from nitrite, nitrate must be reduced to nitrite enzymatically using nitrate reductase (not included) before performing the assay so that the total amount of nitrite can be measured. The kit includes ready-to-use Griess reagent, nitrite standard solution, and a detailed protocol.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a941acf167fb29661515b20e5951bb90d5e232b2&vid=a941acf167fb29661515b20e5951bb90d5e232b2&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df9540d3eb5a5566df70426b4ea44593d02aaf51&vid=df9540d3eb5a5566df70426b4ea44593d02aaf51&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e3d5c87ee4bfc7366604979d538d4a7ba156904&vid=9e3d5c87ee4bfc7366604979d538d4a7ba156904&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d2524b3e7182e82ec8e5bc9663d7c89ecb3066&vid=32d2524b3e7182e82ec8e5bc9663d7c89ecb3066&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e3c41c232637ad240449d1ab135addc2d0d3711&vid=2e3c41c232637ad240449d1ab135addc2d0d3711&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d5561f3a235caaed5c0d7dc9a93c692fc34f6c2&vid=3d5561f3a235caaed5c0d7dc9a93c692fc34f6c2&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1c156c4fd509c7874c41d4a006ec4cab8c8f594&vid=e1c156c4fd509c7874c41d4a006ec4cab8c8f594&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge