8.51030.8500
8.51030.8100
01-63-0134-500EA
409
EUR
InStock
01-63-0134-500
01-63-0134-100
5(6)-TAMRA ≥97% (durch TLC), Novabiochem®
5(6)-TAMRA
5(6)-carboxytetramethylrhodamine (TAMRA; λex555 nm, λem 580 nm) is a useful tool for preparing fluorescently-labeled peptides.
- Supplied as a mixture of 5-and 6-isomers
- Cost effective